Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Human
- Size:20 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening.
- Endotoxin content:<1.0 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:2720
- Protein synonyms:MPS4B|67kDa|EBP|beta 1|elastin receptor 1 (67kD)|Acid beta-galactosidase|Lactase|Elastin receptor 1|betaGalactosidase1|GLB1|ELNR1|beta-galactosidase|EC 3.2.1.23|galactosidase|elastin receptor 1
- Accession no.:AAA51819
- Protein/peptide name:beta-Galactosidase-1/GLB1
- Purity:>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain at 5 ╡g per lane
- Molecular weight:70-95 kDa, reducing conditions
- Cat. no.:102868-580
- Supplier No.:6464-GH-020
The Recombinant Human beta-Galactosidase-1/GLB1 Protein from R&D Systems is derived from CHO. The Recombinant Human beta-Galactosidase-1/GLB1 Protein has been validated for the following applications: Enzyme Activity.