Order Entry
Canada
Orders LinkContactUsLinkComponent
Mouse Recombinant DRAXIN (from NS0 cells)

Mouse Recombinant DRAXIN (from NS0 cells)

25 μg
Catalog # 102867-568
Marketsource
Supplier #:  6149-DR-025
Mouse Recombinant DRAXIN (from NS0 cells)
Catalog # 102867-568
Supplier:  
Marketsource
CAS Number:  
Mouse Recombinant DRAXIN (from NS0 cells)

25 μg

CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

The Recombinant Mouse Draxin Protein from R&D Systems is derived from NS0. The Recombinant Mouse Draxin Protein has been validated for the following applications: Bioactivity.

Specifications

Specifications

  • Size:
    25 μg
  • Protein/peptide name:
    DRAXIN
  • Protein synonyms:
    dorsal inhibitory axon guidance protein|UNQ3119|MGC117222|Dorsal repulsive axon guidance protein|chromosome 1 open reading frame 187|C1orf187|draxin|AGPA3119|FLJ34999|Neucrin
  • Purity:
    >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
  • Accession no.:
    Q6PAL1
  • Source:
    NS0 cells
  • Molecular weight:
    45-60 kDa, reducing conditions
  • Cat. No.:
    102867-568
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.10 EU per 1 ╡g of the protein by the LAL method.
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Gene ID:
    374946
  • Conjugation:
    Unconjugated
  • Species:
    Mouse

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.