Mouse Recombinant Sonic Hedgehog/Shh (from E. coli)
Catalog # 102868-230
Supplier: R&D Systems
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Size:200 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:6469
- Protein synonyms:TPT|HLP3|sonic hedgehog|SMMCIsonic hedgehog homolog (Drosophila)|HHG1|sonic hedgehog protein|HPE3|Shh|sonic hedgehog homolog|TPTPS|HHG-1|MCOPCB5sonic hedgehog (Drosophila) homolog
- Accession no.:Q62226
- Protein/peptide name:Sonic Hedgehog/Shh
- Purity:>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
- Cat. no.:102868-230
- Supplier No.:464-SH-200
Specifications
About this item
The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus from R&D Systems is derived from E. coli. The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus has been validated for the following applications: Bioactivity.