Order Entry
Canada
ContactUsLinkComponent
Human Recombinant TGF-beta 3 (from Baculovirus (Sf21 Insect Cells))
Human Recombinant TGF-beta 3 (from Baculovirus (Sf21 Insect Cells))
Catalog # 102870-988
Supplier:  R&D Systems
CAS Number:  
Human Recombinant TGF-beta 3 (from Baculovirus (Sf21 Insect Cells))
Catalog # 102870-988
Supplier:  R&D Systems
Supplier Number:  243-B3-200
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    Baculovirus (Sf21 Insect cells)
  • Species:
    Human
  • Size:
    200 μg
  • Storage conditions:
    Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
  • Endotoxin content:
    <0.01 EU per 1 ╡g of the protein by the LAL method.
  • Gene ID:
    7043
  • Protein synonyms:
    TGFbeta 3|TGF-beta-3|transforming growth factor|transforming growth factor beta-3|FLJ16571|beta 3|ARVD|TGFB3|TGF-beta3
  • Accession no.:
    P10600
  • Protein/peptide name:
    TGF-beta 3
  • Purity:
    >97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
  • Molecular weight:
    12 kDa, reducing conditions 24 kDa, non-reducing conditions
  • Cat. no.:
    102870-988
  • Supplier No.:
    243-B3-200

Specifications

About this item

The Recombinant Human TGF-beta 3 Protein from R&D Systems is derived from Sf 21 (baculovirus). The Recombinant Human TGF-beta 3 Protein has been validated for the following applications: Bioactivity.