Human Recombinant LDL R (from NS0 cells)
102870-552
:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:NS0 cells
- Species:Human
- Size:25 μg
- Storage conditions:Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
- Endotoxin content:<0.10 EU per 1 ╡g of the protein by the LAL method.
- Gene ID:3949
- Protein synonyms:low-density lipoprotein receptor class A domain-containing protein 3|FHC|LDLR|FH|LDLCQ2|low-density lipoprotein receptor|LDL receptor|low density lipoprotein receptor
- Accession no.:P01130
- Protein/peptide name:LDL R
- Purity:>90%, by SDS-PAGE under reducing conditions and visualized by silver stain
- Molecular weight:120-145 kDa, reducing conditions
- Cat. no.:102870-552
- Supplier No.:2148-HP-025
The Recombinant Human LDL R (High Purity) Protein from R&D Systems is derived from NS0. The Recombinant Human LDL R (High Purity) Protein has been validated for the following applications: Bioactivity.