Order Entry
Canada
ContactUsLinkComponent
StrataClean Resin, Agilent Technologies
StrataClean Resin, Agilent Technologies
Certificates
undefined
StrataClean Resin, Agilent Technologies
Certificates

Specifications

  • Method / Format:
    Affinity resin
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
    Amplified/modified DNA
  • Sample type:
    DNA preps
    PCR and other enzymatic reactions
  • Target molecule:
    DNA
  • Description:
    StrataClean Resin

Specifications

Related Information

Frequently Bought Together