BriskHeat® Adjustable Wrap-Around Tote Tank/IBC Blanket Heaters, Cole Parmer
Supplier: Antyila Scientific
Certificates
About this item
The full coverage plug and play system is constructed of a silicone impregnated cloth facing and liner with a 1/4"-thick fiberglass insulation, and is intended for indoor use.
- Provide fast uniform heat to your tote tank/IBC contents with this wrap-around blanket heater
- Avoid freezing and increase flow rates of tote tank/IBC contents.
- Built-in manual reset high-limit thermostat offers added versatility
- Setup and operation is simple: wrap the blanket around your tote tank, secure it with the adjustable nylon straps, and set temperature
Blanket heater will fit any tote tank width from 40x40" to 48x48".
120 VAC, 1440 W, 12 A
Certifications: CE approved, RoHS compliant.
Accessories information: BriskHeat TOTE-TOP Insulated top cover is sold separately. Compatible with all adjustable wrap-around tote tank/IBC blanket heaters.
Ordering information: Includes 6-ft power cord with standard 3-prong plug.
Caution: For indoor use in non-hazardous areas only.
Frequently Bought Together