Anti-IL-5 Rabbit Polyclonal Antibody (Biotin)
Supplier: Peprotech
Certificates
About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-5. AntiHuman IL-5specific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: IL-5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10781-476
- 10781-474
- Antigen symbol:
- IL-5
- IL-5
- Antigen name:
- Interleukin-5
- Interleukin-5
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human IL-5
- Recombinant Human IL-5
- Form:
- Lyophilized
- Lyophilized
- Molecular weight:
- 26.5 kDa
- 26.5 kDa
- Purification:
- Recombinant Human IL-5
- Recombinant Human IL-5
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together