About this item
Produced from sera of rabbits immunized with highly pure Recombinant Murine Eotaxin (CCL11). AntiMurine Eotaxin (CCL11)specific antibody was purified by affinity chromatography employing an immobilized Murine Eotaxin matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: Eotaxin (CCL11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10775-584
- 10775-586
- 10775-582
- Antigen symbol:
- Eotaxin (CCL11)
- Eotaxin (CCL11)
- Eotaxin (CCL11)
- Antigen name:
- Eotaxin
- Eotaxin
- Eotaxin
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Murine Eotaxin (CCL11)
- Recombinant Murine Eotaxin (CCL11)
- Recombinant Murine Eotaxin (CCL11)
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular weight:
- 8.4 kDa
- 8.4 kDa
- 8.4 kDa
- Purification:
- Recombinant Murine Eotaxin (CCL11)
- Recombinant Murine Eotaxin (CCL11)
- Recombinant Murine Eotaxin (CCL11)
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient