About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human sTNF Receptor Type II. AntiHuman sTNF Receptor Type IIspecific antibody was purified by affinity chromatography employing an immobilized Human sTNF Receptor Type II matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: sTNF Receptor Type
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10776-550
- 10776-554
- 10776-552
- Antigen symbol:
- sTNF Receptor Type
- sTNF Receptor Type
- sTNF Receptor Type
- Antigen name:
- sTNF
- sTNF
- sTNF
- Antigen synonyms:
- Type
- Type
- Type
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Human sTNF Receptor Type II
- Recombinant Human sTNF Receptor Type II
- Recombinant Human sTNF Receptor Type II
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular weight:
- 18.9 kDa
- 18.9 kDa
- 18.9 kDa
- Purification:
- Recombinant Human sTNF Receptor Type II
- Recombinant Human sTNF Receptor Type II
- Recombinant Human sTNF Receptor Type II
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient