About this item
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-6. AntiHuman IL-6specific antibody was purified by affinity chromatography employing an immobilized Human IL-6 matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10781-484
- 10781-482
- 10781-480
- Antigen symbol:
- IL-6
- IL-6
- IL-6
- Antigen name:
- IL-6
- IL-6
- IL-6
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Human IL-6
- Recombinant Human IL-6
- Recombinant Human IL-6
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular weight:
- 20.9 kDa
- 20.9 kDa
- 20.9 kDa
- Purification:
- Recombinant Human IL-6
- Recombinant Human IL-6
- Recombinant Human IL-6
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient