To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen name:IGF-I
- Antigen symbol:IGF-I
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:50 µg
- Form:Lyophilized
- Storage buffer:See Product Data Sheet for more information.
- Molecular weight:7.6 kDa
- Storage temperature:Store at –20 °C prior to reconstitution.
- Shipping temperature:Ambient
- Immunogen:Recombinant Human IGF-I
- Purification:Recombinant Human IGF-I
- Cat. no.:10775-010
- Packaging:PP Vial
- Supplier No.:500P1150UG
Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-I. AntiHuman IGF-Ispecific antibody was purified by affinity chromatography employing an immobilized Human IGF-I matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: IGF-I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human