To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:sTRAIL/Apo2L
- Antigen symbol:sTRAIL/Apo2L
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Size:500 µg
- Form:Lyophilized
- Storage buffer:See Product Data Sheet for more information.
- Molecular weight:19.6 kDa
- Storage temperature:Store at –20 °C prior to reconstitution.
- Shipping temperature:Ambient
- Immunogen:Recombinant Human sTRAIL/Apo2L
- Purification:Recombinant Human sTRAIL/Apo2L
- Cat. no.:10774-884
- Packaging:PP Vial
- Supplier No.:500M49500UG
Specifications
About this item
Monoclonal AntibodyProduced in BALB/c x ICR F1-mice immunized with highly pure Recombinant Human sTRAIL/Apo2L. AntiHuman sTRAIL/Apo2Lspecific antibody was purified from ascites fluid by Protein A affinity chromatography.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: sTRAIL/Apo2L
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human