Anti-NT-4 Goat Polyclonal Antibody (Biotin)
Supplier: Peprotech
Certificates
About this item
Produced from sera of goats immunized with highly pure Recombinant Human NT-4. AntiHuman NT-4specific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: NT-4
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10775-756
- 10775-758
- Antigen symbol:
- NT-4
- NT-4
- Antigen name:
- NT-4
- NT-4
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human NT-4
- Recombinant Human NT-4
- Form:
- Lyophilized
- Lyophilized
- Molecular weight:
- 28.1 kDa
- 28.1 kDa
- Purification:
- Recombinant Human NT-4
- Recombinant Human NT-4
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together