Anti-NAP-2 (CXCL7) Goat Polyclonal Antibody (Biotin)
Supplier: Peprotech
Certificates
About this item
Produced from sera of goats immunized with highly pure Recombinant Human NAP-2 (CXCL7). AntiHuman NAP-2 (CXCL7)specific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: NAP-2 (CXCL7)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Specifications
- Catalog No:
- 10774-964
- 10774-966
- Antigen symbol:
- NAP-2 (CXCL7)
- NAP-2 (CXCL7)
- Antigen name:
- NAP-2
- NAP-2
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human NAP-2 (CXCL7)
- Recombinant Human NAP-2 (CXCL7)
- Form:
- Lyophilized
- Lyophilized
- Molecular weight:
- 7.6 kDa
- 7.6 kDa
- Purification:
- Recombinant Human NAP-2 (CXCL7)
- Recombinant Human NAP-2 (CXCL7)
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together