About this item
Produced from sera of goats immunized with highly pure Recombinant Human MCP-4 (CCL13). AntiHuman MCP-4 (CCL13)specific antibody was purified by affinity chromatography and then biotinylated.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Secondary
Antigen: MCP-4 (CCL13)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10774-990
- 10774-988
- Antigen symbol:
- MCP-4 (CCL13)
- MCP-4 (CCL13)
- Antigen name:
- MCP-4
- MCP-4
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Goat
- Goat
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human MCP-4 (CCL13)
- Recombinant Human MCP-4 (CCL13)
- Form:
- Lyophilized
- Lyophilized
- Molecular weight:
- 8.6 kDa
- 8.6 kDa
- Purification:
- Recombinant Human MCP-4 (CCL13)
- Recombinant Human MCP-4 (CCL13)
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient