About this item
Produced from sera of goats immunized with highly pure Recombinant Human LD78β (CCL3L1). AntiHuman LD78β (CCL3L1)specific antibody was purified by affinity chromatography employing an immobilized Human LD78β matrix.
Applications tested can vary by lot number. See Product Data Sheet for more information.
Type: Primary
Antigen: LD78beta (CCL3L1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10776-724
- 10776-726
- 10776-722
- Antigen symbol:
- CCL3L1
- CCL3L1
- CCL3L1
- Antigen name:
- LD78beta
- LD78beta
- LD78beta
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Human
- Host:
- Goat
- Goat
- Goat
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant Human LD78β (CCL3L1)
- Recombinant Human LD78β (CCL3L1)
- Recombinant Human LD78β (CCL3L1)
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Molecular weight:
- 7.7 kDa
- 7.7 kDa
- 7.7 kDa
- Purification:
- Recombinant Human LD78β (CCL3L1)
- Recombinant Human LD78β (CCL3L1)
- Recombinant Human LD78β (CCL3L1)
- Storage buffer:
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- See Product Data Sheet for more information.
- Storage temperature:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient