Order Entry
Canada
ContactUsLinkComponent
Anti-sFas Ligand Goat Polyclonal Antibody
Anti-sFas Ligand Goat Polyclonal Antibody
Supplier:  Peprotech
Certificates
Anti-sFas Ligand Goat Polyclonal Antibody
Supplier:  Peprotech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Catalog No:
  • 10776-720
  • 10776-718
  • 10771-456
  • Antigen symbol:
  • sFas Ligand
  • sFas Ligand
  • sFas Ligand
  • Antigen name:
  • sFas
  • sFas
  • sFas
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Goat
  • Goat
  • Goat
  • Isotype:
  • IgG
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human sFas Ligand
  • Recombinant Human sFas Ligand
  • Recombinant Human sFas Ligand
  • Form:
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Molecular weight:
  • 0
  • 0
  • 0
  • Purification:
  • Recombinant Human sFas Ligand
  • Recombinant Human sFas Ligand
  • Recombinant Human sFas Ligand
  • Storage buffer:
  • See Product Data Sheet for more information.
  • See Product Data Sheet for more information.
  • See Product Data Sheet for more information.
  • Storage temperature:
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Store at –20 °C prior to reconstitution.
  • Shipping temperature:
  • Ambient
  • Ambient
  • Ambient

Specifications

Frequently Bought Together