Mouse Recombinant TFF-1 (from E. coli)
The Trefoil Factor peptides (TFF1, TFF2 and TFF3) are expressed in the gastrointestinal tract, and appear to play an important role in intestinal mucosal defense and repair. TFF1 is essential for normal differentiation of the antral and pyloric gastric mucosa, and functions as a gastric-specific tumor suppressor gene. Recombinant Murine TFF-1 is a 7.4 kDa monomeric protein consisting of a 66 amino acid polypeptide chain, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds.
The Trefoil Factor peptides (TFF1, TFF2 and TFF3) are expressed in the gastrointestinal tract, and appear to play an important role in intestinal mucosal defense and repair. TFF1 is essential for normal differentiation of the antral and pyloric gastric mucosa, and functions as a gastric-specific tumor suppressor gene. Recombinant Murine TFF-1 is a 7.4 kDa monomeric protein consisting of a 66 amino acid polypeptide chain, which includes a 40-amino acid trefoil motif containing three conserved intramolecular disulfide bonds.
- Catalog No:
- 76077-358
- 76077-360
- 76077-462
- 76077-464
- 76077-466
- 76077-468
- Protein/peptide name:
- TFF-1
- TFF-1
- TFF-1
- TFF-1
- TFF-1
- TFF-1
- Protein synonyms:
- Trefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)
- Trefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)
- Trefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)
- Trefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)
- Trefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)
- Trefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)
- Protein/peptide type:
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Recombinant
- Species:
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Mouse
- Source:
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- E. coli
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Unconjugated
- Gene ID:
- 21784 Q08423
- 21784 Q08423
- 21784 Q08423
- 21784 Q08423
- 21784 Q08423
- 21784 Q08423
- Sequence:
- QAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPF
- QAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPF
- QAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPF
- QAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPF
- QAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPF
- QAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPF
- Protease-free:
- Y
- Y
- Y
- Y
- Y
- Y
- Endotoxin level:
- Low
- Low
- Low
- Low
- Low
- Low
- Endotoxin content:
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- >0.1 ng/μg
- Biological activity:
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Biological Activity tests can vary. Please refer to the Product CoA for information.
- Purity:
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- 0.98
- Form:
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Lyophilized
- Formulation:
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Formulation can vary. Please refer to product CoA for information.
- Molecular weight:
- 7.4 kDa
- 7.4 kDa
- 7.4 kDa
- 7.4 kDa
- 7.4 kDa
- 7.4 kDa
- Reconstitution instructions:
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Reconstitution instructions can vary. Please refer to the product CoA for info.
- Storage conditions:
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Store at –20 °C prior to reconstitution.
- Shipping temperature:
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
- Ambient
Product Family Options
- Size
- : 315-31-500UG
Specifications
Cat. No.76077-358Protein/peptide nameTFF-1Protein synonymsTrefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID21784 Q08423SequenceQAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 315-31-1MG
Specifications
Cat. No.76077-360Protein/peptide nameTFF-1Protein synonymsTrefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID21784 Q08423SequenceQAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 315-31-5UG
Specifications
Cat. No.76077-462Protein/peptide nameTFF-1Protein synonymsTrefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID21784 Q08423SequenceQAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 315-31-20UG
Specifications
Cat. No.76077-464Protein/peptide nameTFF-1Protein synonymsTrefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID21784 Q08423SequenceQAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 315-31-100UG
Specifications
Cat. No.76077-466Protein/peptide nameTFF-1Protein synonymsTrefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID21784 Q08423SequenceQAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient - : 315-31-250UG
Specifications
Cat. No.76077-468Protein/peptide nameTFF-1Protein synonymsTrefoil Factor 1|HP1.A|pS2 protein|PNR2|Breast Cancer Estrogen-Inducible protein (BCEI)Protein/peptide typeRecombinantSpeciesMouseSourceE. coliConjugationUnconjugatedGene ID21784 Q08423SequenceQAQAQAQAQE ETCIMAPRER INCGFPGVTA QQCTERGCCF DDSVRGFPWC FHPMAIENTQ EEECPFProtease-freeYEndotoxin levelLowEndotoxin content>0.1 ng/μgBiological activityBiological Activity tests can vary. Please refer to the Product CoA for information.Purity0.98FormLyophilizedFormulationFormulation can vary. Please refer to product CoA for information.Molecular weight7.4 kDaReconstitution instructionsReconstitution instructions can vary. Please refer to the product CoA for info.Storage conditionsStore at –20 °C prior to reconstitution.Shipping temperatureAmbient
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



