Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant sCD4 (from CHO Cells)
 :  Peprotech
 
Human Recombinant sCD4 (from CHO Cells)
 :  Peprotech
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Unconjugated
  • Protein/peptide type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C prior to reconstitution.
  • Endotoxin content:
    >0.1 ng/μg
  • Biological activity:
    Biological Activity tests can vary. Please refer to the Product CoA for information.
  • Form:
    Lyophilized
  • Gene ID:
    920 P01730
  • Reconstitution instructions:
    Reconstitution instructions can vary. Please refer to the product CoA for info.
  • Protease-free:
    Y
  • Protein synonyms:
    OKT4(a-f)|L3T4|Leu-3(a|b)|T-cell surface antigen T4/Leu-3|T-cell surface glycoprotein CD4|Cluster determinant 4|Interleukin IL-16 receptor
  • Protein/peptide name:
    sCD4
  • Purity:
    0.95
  • Molecular weight:
    41.3 kDa
  • Sequence:
    KKVVLGKKGD TVELTCTASQ KKSIQFHWKN SNQIKILGNQ GSFLTKGPSK LNDRADSRRS LWDQGNFPLI IKNLKIEDSD TYICEVEDQK EEVQLLVFGL TANSDTHLLQ GQSLTLTLES PPGSSPSVQC RSPRGKNIQG GKTLSVSQLE LQDSGTWTCT VLQNQKKVEF KIDIVVLAFQ KASSIVYKKE GEQVEFSFPL AFTVEKLTGS GELWWQAERA SSSKSWITFD LKNKEVSVKR VTQDPKLQMG KKLPLHLTLP QALPQYAGSG NLTLALEAKT GKLHQEVNLV VMRATQLQKN LTCEVWGPTS PKLMLSLKLE NKEAKVSKRE KAVWVLNPEA GMWQCLLSDS GQVLLESNIK VLPTWSTPVQ P
  • Endotoxin level:
    Low
  • Formulation:
    Formulation can vary. Please refer to product CoA for information.
  • Shipping temperature:
    Ambient

 

Frequently Bought Together