Human IL-22 Standard ABTS ELISA Development Kit, PeproTech, Inc.
About this item
PeproTech's Human IL-22 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-22 in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human IL-22 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-22 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human IL-22 in approximately 1000 ELISA plate wells.
1 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
PeproTech's Human IL-22 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-22 in a sandwich ELISA format.
- Easy-to-use
- Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human IL-22 ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-22 in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human IL-22 in approximately 1000 ELISA plate wells.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 10777-898
- 10781-658
- Target protein:
- IL-22
- IL-22
- Primary antibody reactivity:
- Human
- Human
- Assay Type:
- Sandwich
- Sandwich
Specifications
Product Family Options
Product Information
- SizeAvailabilityPrice
- 10777-898Human IL-22 Mini-ELISA KitHuman IL-22 Mini-ELISA KitKit components sufficient to assay 2 platesDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10777-898Target proteinIL-22Primary antibody reactivityHumanAssay TypeSandwich - 10781-658Human IL-22 ELISA KitHuman IL-22 ELISA KitKit components sufficient to assay 10 platesLog in for availabilityKit of 10$1,096.27Supplier Number: 900-K246KItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:
More
Specifications
Cat. No.10781-658Target proteinIL-22Primary antibody reactivityHumanAssay TypeSandwich
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



