Vernier® Go Direct™ Current Sensor
Product Details & Documents
The Go Direct Current Probe measures the electric currents in circuits.
- Capture small currents like those produced by a magnet falling through a coil
- Use in combination with the Go Direct voltage probe to investigate Ohm’s law or series and parallel circuits
- Explore RC and RLC circuits
Simplify your experimental setup with the Go Direct Current Probe. It connects wirelessly via Bluetooth® or wired via USB to your device. The wireless connection eliminates additional cables that can clutter the lab bench.
The Go Direct robust rechargeable battery offers long battery life and provides always-ready operation. Monitor battery life directly from our free Graphical Analysis™ 4 app. Should the battery run low, simply connect the sensor to the charging cable and keep on collecting-no interruptions or inconvenience to you or your students.
Our Go Direct sensors connect directly to your mobile device, Chromebook™, or computer using our free Graphical Analysis™ 4 app-no additional equipment or software purchases are necessary. Go Direct sensors can be used wired via USB or wirelessly via Bluetooth wireless technology, allowing you to choose the solution best for your classroom or laboratory.
Caution: These products are designed for educational use. Our products are not designed nor are they recommended for any industrial, medical, or commercial process such as life support, patient diagnosis, control of a manufacturing process, or industrial testing of any kind.
Specifications
- Description:Go Direct™ current sensor
- Cat. No.:CA470303-840
- Interface:USB