One-Touch Pump, Menda
Catalog #: 15431-758
Supplier: Desco
Specifications
- Description:One-touch pump, without stem
- Cat. no.:15431-758
- Supplier No.:35300
Specifications
About this item
One-Touch pumps are made of precision machined valves for smooth consistent operation that also eliminates spraying and splashing.
- Cam-lock hinge keeps lid open and closed that reduces odors
- Fits all Standard Menda HDPE, glass, and steel bottles
- Dispenses approximately 0.2 cc of liquid with each pump
- Silicone and latex free that are perfect for ultra-clean environments
- Not recommended for use with acrylic liquid
- Made of stainless steel
Customers Who Bought This Also Bought