You searched for: Chromatography Columns
Chromatography columns separate molecules within mixtures during gas or liquid methods. Filled with stationary phase compositions or resins, the prepacked options reduce the preparing time required. The vertical pieces are able to withstand extreme temperatures and pressure fluctuations experienced when the mobile phase is forced through the medium. Meet preparative and analytical laboratory procedures with the chromatography columns to separate dissimilar moving components based on individual polarity, composition, and size.
Avantor® Hichrom HI-20, GC Columns
Supplier: VWR International
General purpose column, ideal as confrmation column.
Expand 156 Items
Avantor® Hichrom HI-13, GC Columns
Supplier: VWR International
General purpose column, ideal as confrmation column.
Expand 156 Items
Avantor® Hichrom HI-SE30, GC Columns
Supplier: VWR International
General purpose apolar column.
Expand 157 Items
Avantor® Hichrom HI-SE54 HT, GC Columns
Supplier: VWR International
High Temperature general purpose column for high boiling petroleum products, Long-chained hydrocarbons.
Expand 24 Items
Avantor® Hichrom HI-WAX Plus, GC Columns
Supplier: VWR International
Max temperature limits may change depending on the film thickness.
Expand 149 Items
Avantor® Hichrom HI-70, GC Columns
Supplier: VWR International
Max temperature limits may change depending on the film thickness.
Expand 36 Items
Avantor® Hichrom HI-35 HT, GC Columns
Supplier: VWR International
General purpose column with added high temperature stability for the analysis of Pesticides, PCBs, substituted polar compounds, phenols, etc.
Expand 23 Items
Avantor® Hichrom HI-17 HT, GC Columns
Supplier: VWR International
High Temperature general purpose column. Ideal for confrmation analysis, ideal as high polarity dimension in GCxGC-HT confgurations.
Expand 23 Items
Avantor® Hichrom HI-ACID (FFAP), GC Columns
Supplier: VWR International
Very popular nitroterephthalic-acid-modified polyethylene glycol (PEG) column of high polarity for the analysis of free fatty acids, phenols, alcohols, glycols.
Expand 149 Items
Avantor® Hichrom HI-DEX G-01, GC Columns
Supplier: VWR International
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Expand 12 Items
Avantor® Hichrom, Retention Gap/Precolumn, HMDS Deactivated Fused Silica
Supplier: VWR International
HMDS deactivated fused silica retention gap / precolumn. Deactivation mainly for apolar solvents.
Expand 48 Items
Avantor® Hichrom HI-VOC 1, GC Columns
Supplier: VWR International
Application specifc columns for volatile organic compounds (VOCs), solvents and purgeable compounds. Due to the high max. temperature, they are ideal for two-parallel columns confg. in the same oven. A good alternative to the HI-624 giving different selectivity. It is a more robust than the HI-624 and can operate at higher temperatures.
Expand 42 Items
Avantor® Hichrom HI-35, GC Columns
Supplier: VWR International
Really versatile general purpose mid-polarity phase for a wide range of applications. Ideal in confirmation analysis.
Expand 145 Items
Avantor® Hichrom HI-FFAP EXT, GC Columns
Supplier: VWR International
Very popular nitroterephthalic-acid-modified polyethylene glycol (PEG) column of high polarity for the analysis of free fatty acids, phenols, alcohols, glycols, with extended temperature range.
Expand 150 Items
Avantor® Hichrom, Retention Gap/Precolumn, DPTMDS Deactivated Fused Silica
Supplier: VWR International
DPTMDS deactivated fused silica retention gap/precolumn. General purpose deactivation for both apolar and polar solvents.
Expand 48 Items
Avantor® Hichrom HI-PLUS 25, GC Columns
Supplier: VWR International
Offering a range of selectivity's with differing copolymer rations of PEG and methyl polysiloxane. HI-PLUS, HI-PLUS 10, HI-PLUS 25, HI-PLUS 75, HI-PLUS 90.
Expand 124 Items
Avantor® Hichrom HI-8 HT, GC Columns
Supplier: VWR International
High Temperature general purpose column ideal for PCBs compounds, Pesticides, environmental analysis.
Expand 24 Items
Avantor® Hichrom HI-DAI 2, GC Columns
Supplier: VWR International
Application specifc column for direct introduction of aqueous samples, thus minimizing sample preparation.
Expand 6 Items
Avantor® Hichrom HI-PAH 2, GC Columns
Supplier: VWR International
Application specifc column for polycyclic aromatics hydrocarbons.
Expand 4 Items
Avantor® Hichrom HI-ALC 1, GC Columns
Supplier: VWR International
Application specific column for blood alcohols testing.
Expand 3 Items
Avantor® Hichrom HI-SOLVE 1, GC Columns
Supplier: VWR International
Application specifc columns developed for complex solvents mixtures analysis. HI-SOLVE 1 is ideal for aromatics and oxygenates in gasoline.
Expand 1 Items
Avantor® Hichrom HI-SE54, GC Columns
Supplier: VWR International
General purpose column for solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, allergens, pesticides, and so on.
Expand 165 Items
Avantor® Hichrom, Retention Gap/Precolumn, CW Deactivated Fused Silica
Supplier: VWR International
CW deactivated fused silica retention gap / precolumn. Deactivation mainly for polar solvents.
Expand 48 Items
Avantor® Hichrom HI-WAX BA, GC Columns
Supplier: VWR International
These columns maximum temperature limits may change depending on the film thickness.
Expand 150 Items
Avantor® Hichrom HI-1301, GC Columns
Supplier: VWR International
Max temperature limits may change depending on the film thickness.
Expand 146 Items
Avantor® Hichrom HI-OVW, GC Columns
Supplier: VWR International
Max temperature limits may change depending on the film thickness.
Expand 1 Items
Avantor® Hichrom HI-5 HT, GC Columns
Supplier: VWR International
General purpose column with added temperature stability. For the analysis of solvent impurities, PCBs, hydrocarbons, essential oils, semivolatiles, pesticides, phenols and more.
Expand 24 Items
Avantor® Hichrom HI-DEX B-SE, GC Columns
Supplier: VWR International
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Expand 12 Items
Avantor® Hichrom HI-DEX DAC Gamma, GC Columns
Supplier: VWR International
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Expand 12 Items
Avantor® Hichrom HI-DEX DET Gamma, GC Columns
Supplier: VWR International
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.