Orders LinkContactUsLinkComponent
Caspar Screwdriver, Sklar
Caspar Screwdriver, Sklar
Caspar Screwdriver, Stainless steel, 7⅞"
Catalog # 76191-480
Supplier:   Sklar
Supplier #:  13-7052
Caspar Screwdriver, Sklar
Catalog # 76191-480
Supplier:  Sklar
Caspar Screwdriver, Sklar
Caspar Screwdriver, Sklar
Caspar Screwdriver, Stainless steel, 7⅞"
Price
$1,636.21
New Customer Promotional Price
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Sklar products are industry standard.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
Sklar's® Caspar Screwdriver is for use with Caspar distraction screws to hold vertebral implant plates in place. It is a pentagonal shaped screwdriver that fits over part of the screw versus simply into the tip. It has an overall length of 8".
Specifications
  • Description:
    Caspar Screwdriver
  • Length:
    7⅞"
  • Material:
    Stainless steel
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)