ContactUsLinkComponent
Marcy® Pulp Density and Specific Gravity Scale, Metso
Marcy® Pulp Density and Specific Gravity Scale, Metso
Catalog # CA11019-902
undefined
Marcy® Pulp Density and Specific Gravity Scale, Metso
Catalog # CA11019-902
Supplier Number:  777005444
Specifications
  • Description:
    Marcy® Pulp Density and Specific Gravity Scale, 10" O-Ø
  • Cat. no.:
    CA11019-902
  • Supplier No.:
    MR125
Specifications
About this item
Direct reading scale determines specific gravities of pulps, liquids and dry solids. This metal gravity scale has a plastic covering over the dial which does not effect the accuracy of the scale. It eliminates errors in reading charts or graphs and prevents errors in calculations.
  • Circular dial is equivalent to a 47" beam balance for greater sensitivity
  • Entire unit is supported by a top ring, scale is accurately set at the factory
  • Cylindrical containers with slotted overflow holes, permit accurate filling to exactly 1000 cc
  • Samples do not have to be weighed separately, refer to tables or make calculations
Interchangeable dials make weighing applicable for any range of solids with light or heavy density. Each scale has 12 interchangeable dial faces with a specific gravity range of dry solids from 1.2 through 7.8.
Use to find weight of sample in grams or kilograms, specific gravity of liquid or pulp, percent solids contained in the pulp of any given specific gravity and specific gravity of dry.
10 lbs. shipping weight, approximate and 3½×12×12" shipping dimensions.