ContactUsLinkComponent
tissueTUBE Sample Tubes with Plugs, Covaris®
tissueTUBE Sample Tubes with Plugs, Covaris®
Catalog # MSPP-520006
Supplier:  COVARIS INC MS
tissueTUBE Sample Tubes with Plugs, Covaris®
Catalog # MSPP-520006
Supplier:  COVARIS INC MS
Supplier Number:  520006
Specifications
  • Description:
    Plugs
  • Cat. no.:
    MSPP-520006
  • For Use With:
    tissueTUBE TT1
  • Supplier No.:
    520006
Specifications
About this item
These tubes are suitable for sample collection, storage, pulverization, and transfer.
  • Ideal for samples of mass 1 g
Used with the CP02 cryoprep pulverizer and with holder tissueTUBE TT1 and tissueTUBE extra thick.