Compartmented Containers for Gel Strips, GenHunter
Catalog # MSPP-B135
Supplier: GENHUNTER CORPORATION MS
Specifications
- Color:Black
- Description:6-Section Container
- Dimensions:101/2L×63/16W×13/4H" (26.7×15.8×4.4 cm) (Bo×); 63/16L×19/16W×13/4H" (15.8×4×4.4 cm) (Compartments)
- Cat. no.:MSPP-B135
- Supplier No.:B135
Specifications
About this item
Containers feature six long sections for longer strips of protein gels.