If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:MiniARCO® Cordless Trimmer Kit
- Cat. no.:MSPP-023326
- Supplier No.:023326
Specifications
About this item
MiniARCO® superior trimmer is an unsurpassed rotary motor direct drive trimmer. This features with a superior quick detachable blade set.
- Powerful rotary motor runs at 6000 rpm
- Compact size, light weight, quiet and cool running
- Continuous run time of up to 45 minutes
The blade is made of precision ground, high carbon steel and with hard chrome finish which resist the rust and corrosion.