Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® TraceableGO™ Bluetooth Data Logging Thermometer

VWR® TraceableGO™ Bluetooth Data Logging Thermometer

VWR® TraceableGO™ Bluetooth datalogging thermometer, T: –20…+70 °C (–4…+158 °F), T: 0.1 °C, T: ±0.4 °C (–10…+70 °C); otherwise ±0.5 °C
Catalog # 76214-376
Supplier:   VWR International
VWR® TraceableGO™ Bluetooth Data Logging Thermometer
Catalog # 76214-376
Supplier:  VWR International
VWR® TraceableGO™ Bluetooth Data Logging Thermometer
VWR® TraceableGO™ Bluetooth Data Logging Thermometer

VWR® TraceableGO™ Bluetooth datalogging thermometer, T: –20…+70 °C (–4…+158 °F), T: 0.1 °C, T: ±0.4 °C (–10…+70 °C); otherwise ±0.5 °C

Price
$227.36
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

A Bluetooth device is the ideal transit partner for monitoring materials between destinations. Safely transport reagents, clinical samples, vaccines, pharmaceuticals, and food items to any location.

  • Press START to begin trip monitoring
  • Place device inside a small cooler or even an entire pallet of material
  • Once the trip has ended, double-press START/STOP to enable Bluetooth and export data using the TraceableGO™ mobile app
  • TraceableGO™ mobile app immediately identifies if the contents experienced conditions outside user-defined parameters
  • Critical information(Temperature, trip duration, MIN/MAX, battery level and alarms) all displayed on LCD at all times
  • Compatible with TraceableGO or TraceableLIVE mobile apps

With the TraceableGO™ mobile app, view if an alarm was triggered, and generate an instant data report. Device stores up to 90 alarm events, records readings once every five minutes by default, or the user may adjust the reading interval between one minute to once every 12 hours. Logging duration is 227 days (7.5 months). TraceableGO™ Bluetooth devices are pre-programmed with alarm values.

Bluetooth data transfer has broad compatibility easy bluetooth connection works with any bluetooth-enabled devices. Once TraceableGO™ arrives, enable the Bluetooth transfer to a smartphone, open the TraceableGO™ app (available on iTunes or Google Play) pair the device to smartphone download the recorded data since the device was enabled and send a report directly from a mobile device.

Certifications: CE marked. RoHS compliant. Traceable to NIST for accuracy. Individually-numbered Traceable® Certificate assures accuracy from an ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National institute of standards and technology). 21 CFR Part 11 compliance with TraceableLIVE App.

Accessories information: Data can be downloaded and devices reconfigured using the free TraceableGO™ app. Optional service allows an unlimited amount of stored data from an unlimited number of TraceableGO™ devices on a TraceableLIVE® account for $30/month.

Ordering information: Supplied with battery and Traceable® certificate.

Specifications
  • Resolution:
    T: 0.1 °C
  • Description:
    VWR® TraceableGO™ Bluetooth datalogging thermometer
  • Accuracy:
    T: ±0.4 °C (–10…+70 °C); otherwise ±0.5 °C
  • Range:
    T: –20…+70 °C (–4…+158 °F)
  • Cat. No.:
    76214-376
  • Recording intervals:
    1 min - 12 h
  • Data memory - No. of readings:
    65536
  • W×D×H:
    89×51×20 mm (3.5×2×0.8")
  • Batteries:
    2 AAA Alkaline batteries (3.0 V)
  • Interface:
    Bluetooth
  • Weight:
    272 g (0.65 oz.)
  • Display:
    LCD
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.