Order Entry
Canada
ContactUsLinkComponent
Pluronic® F-127, 10% Solution in Water, Anaspec
Pluronic® F-127, 10% Solution in Water, Anaspec
Catalog # 103011-196
Supplier:  Anaspec
CAS Number:  
Pluronic® F-127, 10% Solution in Water, Anaspec
Catalog # 103011-196
Supplier:  Anaspec
Supplier Number:  AS-84042
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 mL
  • Storage conditions:
    4°C protected from light
  • Shipping conditions:
    RT
  • Solvent:
    N/A
  • Cat. no.:
    103011-196
  • Supplier No.:
    AS-84042

Specifications

About this item

Cell culture reagent for dissolving AM esters.