Order Entry
Canada
ContactUsLinkComponent
Quinine sulfate Dihydrate *Fluorescence Reference Standard, Anaspec Inc.
Quinine sulfate Dihydrate *Fluorescence Reference Standard, Anaspec Inc.
Catalog # 103010-740
Supplier:  Anaspec
CAS Number:  
Quinine sulfate Dihydrate *Fluorescence Reference Standard, Anaspec Inc.
Catalog # 103010-740
Supplier:  Anaspec
Supplier Number:  AS-80040
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Quinine sulfate
  • Size:
    100 mg
  • Storage conditions:
    -20°C desiccated and protected from light
  • Shipping conditions:
    RT
  • Cat. no.:
    103010-740
  • Absorbency:
    Abs/Em = 348/455 nm
  • Supplier No.:
    AS-80040

Specifications

About this item

Reference standard for measuring fluorescence quantum yield.

Quinine sulfate in 0.5 M H2SO4 has a molar extinction coefficient of 5,700 M-1cm-1 at 347.5 nm. Quinine in acidic media has been widely recommended as a standard for static fluorescence yield determinations. Quinine is less applicable to time-resolved studies, because its fluorescence decay is not monophasic. This is the fluorescence emission spectrum of Quinine sulfate dissolved in 0.5 M H2SO4. The spectrum was taken using an excitation wavelength of 310 nm. The quantum yield is 0.546.

Resuspension condition: Water