To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Secondary
- Antigen name:Immunoglobulin G
- Clonality:Monoclonal
- Clone:Anti-Human/HP6069
- Host:Mouse
- Reactivity:Human
- Antigen symbol:IgG
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Size:100 µg
- Western blot:Yes
- Format:purified liquid
- Form:Liquid
- Storage buffer:pH:
- Storage temperature:Store at 2 - 8°C
- Shipping temperature:4C
- Cat. no.:IC08634521
Specifications
About this item
This is a mouse monoclonal antibody, isotype IgG1, reacting with human IgG1 (Fc Fragment) specific and
conjugated with horseradish peroxidase. No preservative has been added.
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: Anti-Human/HP6069
Conjugation: Horseradish Peroxidase (HRP)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human