Order Entry
Canada
ContactUsLinkComponent
Rhod - 2 AM ≥90% (by HPLC), Ultra Pure Grade
Rhod - 2 AM ≥90% (by HPLC), Ultra Pure Grade
Catalog # 103011-186
Supplier:  Anaspec
CAS Number:  
Rhod - 2 AM ≥90% (by HPLC), Ultra Pure Grade
Catalog # 103011-186
Supplier:  Anaspec
Supplier Number:  AS-84035
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 mg
  • Cat. no.:
    103011-186
  • Supplier No.:
    AS-84035

Specifications

About this item

 :
 :1123.94 g/mol
Storage Temperature:Freezer
CAS Number:129787-64-0
Formula:C₅₂H₅₉BrN₄O₁₉