Order Entry
Canada
ContactUsLinkComponent
Collagen (Type I) (Soluble)
Collagen (Type I) (Soluble)
Catalog # 103011-288
Supplier:  Anaspec
CAS Number:  
Collagen (Type I) (Soluble)
Catalog # 103011-288
Supplier:  Anaspec
Supplier Number:  AS-85102
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    5 mg
  • Storage conditions:
    –20 °C desiccated and protected from light
  • Protein/peptide name:
    Collagen (Type I) (soluble)
  • Shipping temperature:
    Shipped in Blue Ice
  • Cat. no.:
    103011-288
  • Supplier No.:
    AS-85102

Specifications

About this item

Besides water-soluble collagen (Type I) conjugate (Cat#85111), AnaSpec also offers water-insoluble fluoresceinated collagen (type I). It can also be used for fluorometric measurement of collagenase activity. Cat#85102 is the water-insolulbe collagen that is heavily labeled with FITC, resulting in almost total quenching of the conjugates fluorescence. Protease-catalyzed hydrolysis slowly yields brightly green fluorescent dye-labeled peptides. The increase in fluorescence intensity is directly proportional to protease activity. Although this fluoresceinated collagen is more slowly digested by MMP-1 than water-soluble collagen (Cat#85111), it is more specific for the enzyme.

  • Absorbance: Abs/Em = 492/515 nm