Order Entry
Canada
ContactUsLinkComponent
R - PE
R - PE
Catalog # 103011-090
Supplier:  Anaspec Inc
CAS Number:  
R - PE
Catalog # 103011-090
Supplier:  Anaspec Inc
Supplier Number:  AS-82004
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 mg
  • Storage conditions:
    4 °C
  • Protein/peptide name:
    R - PE
  • Formulation:
    in water
  • Shipping temperature:
    Shipped in Blue Ice
  • Cat. no.:
    103011-090
  • Supplier No.:
    AS-82004

Specifications

About this item

R-Phycoerythrin (R-PE), a fluorescent protein from phycobiliprotein family, is isolated from red algae. Its primary absorption peak is at 565 nm with secondary peaks at 496 and 545 nm. The broad excitation spectrum provides the advantage for multi-color immunofluorescent staining or cell sorting. R-PE consists of a, b and g subunits and is present as (ab)6g. R-PE and the closely related BPE are the most intensely fluorescent phycobiliproteins having orange fluorescence. They are significantly brighter and more photostable than conventional organic fluorophores.

  • Absorbance: Abs/Em = 565/575 nm