Order Entry
Canada
ContactUsLinkComponent
CL - APC (Cross Linked - Allophycocyanin)
CL - APC (Cross Linked - Allophycocyanin)
Catalog # 103011-086
Supplier:  Anaspec Inc
CAS Number:  
CL - APC (Cross Linked - Allophycocyanin)
Catalog # 103011-086
Supplier:  Anaspec Inc
Supplier Number:  AS-82002
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 mg
  • Storage conditions:
    4 °C
  • Protein/peptide name:
    CL - APC (Cross Linked - Allophycocyanin)
  • Formulation:
    in water
  • Shipping temperature:
    Shipped in Blue Ice
  • Cat. no.:
    103011-086
  • Supplier No.:
    AS-82002

Specifications

About this item

Allophycocyanin (APC), highly fluorescent phycobiliprotein, is made up of alpha and beta subunits and is present as a trimer ( αβ)3, which is unstable and susceptible to dissociation at low concentrations. The monomer, αβ, has a lower fluorescence quantum yield compared to the trimer and the maximal absorption is also shifted to 620 nm. The chemically cross-linked APC trimer is much more stable than the native APC trimer, but still retains the same spectroscopic properties as the native APC trimer. APC labeled streptavidin, primary and secondary antibodies have been widely used in applications such as flow cytometry, live cell staining, and immunofluorescent staining.

  • Absorbance: Abs/Em = 651/662 nm