Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Microscale Protein Labeling Kit AnaTag™ AMCA - X 'Ultra Convenient'
- Size:1 kit
- Storage conditions:4°C
- Shipping conditions:20
- Cat. no.:103010-368
- Abs/Em:353/442 nm
- Supplier No.:AS-72056
Specifications
About this item
AMCA-X, SE is one of the most popular blue fluorescent tagging molecules. It is widely used to label antibodies, proteins and small drug molecules. AMCA-X succinimidyl ester contains a seven-atom aminohexanoyl spacer between the fluorophore and the reactive group.
This spacer separates the fluorophore from the biomolecule to which it is conjugated, potentially reducing the quenching that typically occurs upon conjugation. AMCA fluorescence can be detected at the emission wavelength of 442±10 nm when excited at 353±10 nm. The AMCA-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all essential components for performing the conjugation reaction and for purifying the AMCA-protein conjugates. The kit is able to perform three conjugation reactions with each reaction labeling up to 200 µg IgG. Kit Size: 3 labeling reactions (3 x 200 µg protein)