To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Fluor 555 Microscale Protein Labeling Kit AnaTag™ HiLyte™ 'Ultra Convenient'
- Size:1 kit
- Storage conditions:4°C
- Cat. no.:103010-352
- Abs/Em:553/568 nm
- Supplier No.:AS-72046
Specifications
About this item
HiLyte™ Fluor 555 acid, SE is an excellent amine-reactive fluorescent labeling dye that generates the protein conjugates that are only slightly red-shifted compared to those of Cy™ 3 dyes, resulting in an optimal match to filters designed for CyTM dyes.
The HiLyte™ Fluor 555-protein conjugates are very stable and suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. Additionally, our in-house data indicated that HiLyte™ Fluor 555 is superior to CyTM 3 in fluorescence polarization-based assays. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 555-protein conjugate. The entire process takes only half an hour. The kit is able to perform three conjugation reactions with each reaction labeling up to 200 µg IgG. Kit Size: 3 labeling reactions (3 x 200 µg protein)