Order Entry
Canada
ContactUsLinkComponent
AnaTag™ HiLyte™ Fluor 488 Protein Labeling Kit [3 labeling reactions (3×5 mg protein)], Anaspec
AnaTag™ HiLyte™ Fluor 488 Protein Labeling Kit [3 labeling reactions (3×5 mg protein)], Anaspec
Catalog # 103010-354
Supplier:  Anaspec Inc
CAS Number:  
AnaTag™ HiLyte™ Fluor 488 Protein Labeling Kit [3 labeling reactions (3×5 mg protein)], Anaspec
Catalog # 103010-354
Supplier:  Anaspec Inc
Supplier Number:  AS-72047
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Fluor 488 Protein Labeling Kit [3 labeling Reactions (3 x 5 mg Protein)] AnaTag™ HiLyte™
  • Size:
    1 kit
  • Storage conditions:
    4°C
  • Shipping conditions:
    20
  • Cat. no.:
    103010-354
  • Abs/Em:
    499/523 nm
  • Supplier No.:
    AS-72047

Specifications

About this item

HiLyte™ Fluor 488, SE is an excellent amine-reactive fluorescent labeling dye for generating protein conjugates. The spectrum of HiLyte™ Fluor488 is similar to fluorescein (FITC), resulting in an optimal match to filters designed for fluorescein.

HiLyte™ Fluor 488 is more photostable than FITC, providing researchers with additional time for capturing image. Its fluorescence can be detected at the emission wavelength of 523±10 nm when excited at 499±10 nm. The HiLyte™ Fluor 488-protein conjugates are very stable and suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 488-protein conjugate. The entire process takes only half an hour. The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein)