MastAssure™ Bacterial Agglutinating Antisera
CA77623-676
New Product
:
- Description:Enterocolitica O group, O-5
- Cat. no.:CA77623-676
- Volume:2 ml
- Details:Factor O-5
An extensive range containing both polyvalent and monovalent diagnostic antisera for the identification of clinical bacteria according to the expression of somatic antigens.
- High quality performance
- Extensive range available
- Easy to read
- Reliable identification
Bacterial serotype can be determined by both slide agglutination and test tube agglutination methods.
The antisera are derived from immunised rabbits, adsorbed to remove cross agglutinins and filter sterilized.