Septa for Autosampler Waste and Wash Vials, PerkinElmer
Catalog # CA97013-074
Supplier: PerkinElmer
Specifications
- Description:Septa for use with viscous or toxic samples
- Cat. no.:CA97013-074
- Supplier No.:N9302780
Specifications
About this item
Silicone septa, 13 mm for screw top waste and wash vials.
Packaging: 50/pack.