Order Entry
Canada
Orders LinkContactUsLinkComponent
Precision Count Beads™, Biolegend®

Precision Count Beads™, Biolegend®

Precision Count Beads™, Supplied in a solution containing 0.02% sodium azide and 0.01% NP40, 100
Catalog # CA75833-874
Supplier:   Biolegend
Supplier #:  424902
Precision Count Beads™, Biolegend®
Catalog # CA75833-874
Supplier:  Biolegend
CAS Number:  
Precision Count Beads™, Biolegend®
Precision Count Beads™, Biolegend®

Precision Count Beads™, Supplied in a solution containing 0.02% sodium azide and 0.01% NP40, 100

CAS Number:  
Price
$432.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Precision Count Beads™ are fluorescent particles about 10 µm that can be detected across a broad range of wavelengths (405 - 635 nm excitation and 400 - 800 nm emission). Absolute numbers of cells can be determined by comparing the bead count and cell count.

Can be used to obtain absolute counts of cells or any other samples acquired on a flow cytometer and also suitable to work with all kinds of assays including a lyse, no-wash whole blood assay.

Specifications

Specifications

  • Description:
    Precision Count Beads™
  • Contents:
    Supplied in a solution containing 0.02% sodium azide and 0.01% NP40
  • No. of tests:
    100
  • Cat. No.:
    CA75833-874
  • Storage conditions:
    2...8 °C

Specifications