Vesta-Syde® SQ st Quaternary Ammonium Disinfectants
Supplier: Steris
Certificates
About this item
STERIS' Vesta-Syde® st SQ quaternary ammonium disinfectant is a liquid concentrate specifically formulated for disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
- Flexibility - Excellent fungicidal efficacy can support reduced sporicide use to control mold contamination
- Convenience - Gamma irradiated and double bagged for easy introduction into your aseptic areas
- Productivity - Formulated with biodegradable surfactants that provide excellent cleaning ability
- Availability - Active Ingredients notified according to the Biocide Product Directive and registered with the US EPA
- Assurance - Each lot documented by a Certificate of Analysis including irradiation and sterility results
STERIS’ Vesta-Syde® SQ st quaternary ammonium disinfectant is the sterile version of the Vesta-Syde® SQ family of disinfectants made specifically for use on hard non-porous surfaces.
The Vesta-Syde® SQ products are all made with the same quaternary ammonium formulation, but are processed and packaged differently for specific applications. At a use-dilution rate of 0.8% v/v (1:128), the Vesta-Syde® SQ products kill a broad spectrum of common bacteria and fungi without harming the surface.
Frequently Bought Together