ContactUsLinkComponent
Waste and Wash Vials for Autosampler, PerkinElmer
  CA97012-220
undefined
Waste and Wash Vials for Autosampler, PerkinElmer
  CA97012-220
 :  09923031
  • Cap included:
    No
  • Capacity:
    4.00 ml
  • Color:
    Clear
  • Description:
    Waste and Wash Vial
  • Cat. no.:
    CA97012-220
  • Supplier No.:
    09923031
These vials are manufactured from Type I Borosilicate Glass, which meets all USP, JP, and EP Pharmacopeia requirements.
  • Excellent at high temperatures
  • Chemical resistant to acidic, neutral and alkali solutions
All vials are packed in a clean environment to ensure you receive contaminant free product every time.
Cap