To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Uric Acid Assay Kit
- Cat. no.:75817-132
- Method:Fluorometric
- Includes:Assay Buffer, Fluorometric Detector, Enzyme Mixture, Assay Reagent, Standard, Plate, and Cover Sheet.
- Supplier No.:700320-96
Specifications
About this item
A fluorescence-based method for detecting uric acid in serum, plasma, and urine.
- Measure uric acid in serum, plasma, and urine
- Assay 40 samples in duplicate
- Assay Range: 0.5 to 10 μM
- Plate-based fluorometric measurement (ex 530 to 540 nm, em 585 to 595 nm)
Uric acid (urate) is the end product of human purine metabolism and is mainly excreted in urine. Many factors, including genetic components and acquired factors, such as obesity and alcohol consumption, influence serum uric acid concentrations. Hyperuricemia induces or facilitates gout, kidney stones, metabolic syndrome, hypertension, and renal and cardiovascular disease, while exercise-induced acute renal failure is a significant complication of renal hypouricemia.