Disposable Tips, MCA 96 nested, for MultiChannel Arm™, Tecan
Catalog # MSPP-30038620
Supplier: TECAN US INC MS
Specifications
- Volume:200 µl
- Cat. no.:MSPP-30038620
- Tip Style:Tecan pure disposable tips, nested for Te-Stack™
- For Use With:MultiChannel Arm™
- Supplier No.:30038620
Specifications
About this item
These disposable tips are used for MultiChannel Arm 96 option for Freedom EVO® liquid handling work stations.
- Improved throughput and walk away time
- Increased flexibility on the work deck, due to SBS format
- Environmentally friendly
- Cost effective
Manufactured, tested and certified to guarantee reliable quality to acquire optimal results.
Tecan Pure: Free from human DNA, DNase, RNase and PCR inhibitors; targeted towards molecular diagnostics, forensics and genomics applications.