Order Entry
Canada
ContactUsLinkComponent
Ammonium acetate ≥99.0% (by anhydrous basis) for HPLC, Fluka™
Ammonium acetate ≥99.0% (by anhydrous basis) for HPLC, Fluka™
Catalog # BJ17836-250G
CAS Number:  
Ammonium acetate ≥99.0% (by anhydrous basis) for HPLC, Fluka™
Catalog # BJ17836-250G
Supplier Number:  17836-250G
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    250 g
  • Cat. no.:
    BJ17836-250G
  • Supplier No.:
    17836-250G

Specifications

Specification Test Results

  • Form:
    Solid
  • assay:
    68.0 - 69.0 %
  • silver (Ag):
    Max. 10 ppb
  • aluminium (Al):
    Max. 20 ppb
  • gold (Au):
    Max. 10 ppb
  • arsenic (As):
    Max. 5 ppb
  • boron (B):
    Max. 10 ppb
  • barium (Ba):
    Max. 10 ppb
  • beryllium (Be):
    Max. 10 ppb
  • bismuth (Bi):
    Max. 10 ppb
  • calcium (Ca):
    Max. 30 ppb
  • cadmium (Cd):
    Max. 10 ppb
  • cobalt (Co):
    Max. 10 ppb
  • chromium (Cr):
    Max. 30 ppb
  • copper (Cu):
    Max. 10 ppb
  • iron (Fe):
    Max. 100 ppb
  • gallium (Ga):
    Max. 10 ppb
  • germanium (Ge):
    Max. 10 ppb
  • indium (In):
    Max. 10 ppb
  • potassium (K):
    Max. 50 ppb
  • lithium (Li):
    Max. 10 ppb
  • magnesium (Mg):
    Max. 50 ppb
  • manganese (Mn):
    Max. 10 ppb
  • molybdenum (Mo):
    Max. 10 ppb
  • sodium (Na):
    Max. 50 ppb
  • nickel (Ni):
    Max. 30 ppb
  • lead (Pb):
    Max. 10 ppb
  • platinum (Pt):
    Max. 10 ppb
  • antimony (Sb):
    Max. 5 ppb
  • tin (Sn):
    Max. 10 ppb
  • strontium (Sr):
    Max. 10 ppb
  • titanium (Ti):
    Max. 50 ppb
  • thallium (Tl):
    Max. 10 ppb
  • vanadium (V):
    Max. 10 ppb
  • zinc (Zn):
    Max. 10 ppb
  • zirconium (Zr):
    Max. 10 ppb
  • chloride (Cl):
    Max. 0.5 ppm
  • phosphate (PO4):
    Max. 0.5 ppm
  • sulfate (SO4):
    Max. 0.5 ppm

Specification Test Results

About this item

 :
 :77.08 g/mol
Merck Index:13,00495
Melting Pt:113 °C
Density:1.170 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-162-9
CAS Number:631-61-8
Formula:H₃CCOONH₄
MDL Number:MFCD00013066