Order Entry
Canada
ContactUsLinkComponent
CHROMABOND® XTR, SPE Columns, Macherey-Nagel
CHROMABOND® XTR, SPE Columns, Macherey-Nagel
Catalog # 29444-586
Supplier:  Macherey-Nagel
Small Business Enterprise
undefined
CHROMABOND® XTR, SPE Columns, Macherey-Nagel
Catalog # 29444-586
Supplier:  Macherey-Nagel
Supplier Number:  730507
Small Business Enterprise

Specifications

  • Capacity:
    70 ml
  • pH range:
    1 - 13
  • Bed Weight:
    14.5 g
  • Column Hardware:
    PP
  • Cat. no.:
    29444-586
  • Phase:
    CHROMABOND® XTR
  • Supplier No.:
    730507

Specifications

About this item

SPE columns, CHROMABOND® XTR, coarse-grained kieselguhr base material with large pore size, high pore volume and constantly high batch-to-batch quality.

  • Fast, reproducible and economical
  • Simultaneous preparation of several samples
  • No problems with phase separation and formation of emulsions
  • High recovery rates, saving of time and solvents
  • Organic solutions need not to be dried after separation

Suitable for liquid-liquid extraction of highly viscous aqueous solutions such as physiological fluids (blood, plasma, and serum) in clinical chemistry, dyes in textiles, environmental and food analysis without use of a separation funnel. High water loadability without breakthrough of water during elution with organic solvents. Also suited for removing small amounts of water from solvents which are not miscible with water.