Phosgene Colorimetric Gas Detector Tubes, Sensidyne
Catalog #: 13502-666
Supplier: Sensidyne
Small Business Enterprise
Specifications
- Measurement range (°Brix):0.5 – 20 ppm (100 mL); 0.1 – 4.0 ppm (500 mL)
- Measurement range:0.5 – 20 ppm (100 mL); 0.1 – 4.0 ppm (500 mL)
- Description:Phosgene Detection Tubes
- Cat. no.:13502-666
- No. of Strokes:1 (100 ml), 5 (500 ml)
- Supplier No.:146S
Specifications
About this item
Phosgene colorimetric gas detector tubes for detection of airborne gases and vapors with detector tube pump and an antibacterial grip.
- Easy to use
- Color changes white to red
- Direct reading from the scale calibrated by 1 pump stroke
- Low-cost method of routine or spot sampling for gases and vapors
Detector tubes safely allow users to perform field screening and tests for the presence of hazardous and toxic gases, and vapors.
Customers Who Bought This Also Bought